missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Acridine-9-carboxylic acid hydrate, 97%

Manufacturer:  Alfa Aesar™ L1079703

 View more versions of this product

Catalog No. AAL1079703



Specifications

Specifications

Acridine-9-carboxylic acid hydrate
1g
C14H11NO3
171327
ZQAZWHSZYVXGMP-UHFFFAOYSA-N
acridine-9-carboxylic acid;hydrate
16211687
97%
~290°C
332927-03-4
MFCD00009734
9-acridinecarboxylic acid hydrate, acridine-9-carboxylic acid hydrate, acmc-1afs1, c14h9no2.h2o, acridin-9-carboxylic acid hydrate, 9-acridinecarboxylic acid hydrate, acridine-9-carboxylic acid hydrate 1:x
C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)C(=O)O.O
241.246
223.23 (Anhydrous)

Chemical Identifiers

332927-03-4
241.246
ZQAZWHSZYVXGMP-UHFFFAOYSA-N
16211687
C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)C(=O)O.O
C14H11NO3
MFCD00009734
9-acridinecarboxylic acid hydrate, acridine-9-carboxylic acid hydrate, acmc-1afs1, c14h9no2.h2o, acridin-9-carboxylic acid hydrate, 9-acridinecarboxylic acid hydrate, acridine-9-carboxylic acid hydrate 1:x
acridine-9-carboxylic acid;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents