Please login to your online account to display your discounted pricing

ACPT-II, Tocris

Competitive mGlu receptor antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material ACPT-II
Assay Percent Range >98%
Molecular Formula C8H11NO6
Formula Weight 217.18
Synonym (1R,3R,4S)-1-Aminocyclopentane-1,3,4-tricarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

111250

 
r & d systems
RDS111250
50mg Each for $1,050.42

111210

 
r & d systems
RDS111210
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 217.18
Quantity 50mg
Chemical Name or Material ACPT-II
Assay Percent Range >98%
Molecular Formula C8H11NO6
CAS 195209-04-2
Synonym (1R,3R,4S)-1-Aminocyclopentane-1,3,4-tricarboxylic acid
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C