Please login to your online account to display your discounted pricing

ACPT-II, Tocris


Competitive mGlu receptor antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS111210

 View more versions of this product

Catalog No. 111210

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 217.18
Quantity 10mg
Chemical Name or Material ACPT-II
Assay Percent Range >98%
Molecular Formula C8H11NO6
CAS 195209-04-2
Synonym (1R,3R,4S)-1-Aminocyclopentane-1,3,4-tricarboxylic acid
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material ACPT-II
Assay Percent Range >98%
Molecular Formula C8H11NO6
Formula Weight 217.18
Synonym (1R,3R,4S)-1-Aminocyclopentane-1,3,4-tricarboxylic acid
Accessories

Desiccate at -20°C