Please login to your online account to display your discounted pricing

ACPT-I, Tocris


Group III mGlu agonist

Quantity 50mg

Manufacturer: r & d systems  RDS111150

 View more versions of this product

Catalog No. 111150

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 217.18
Quantity 50mg
CAS 194918-76-8
Chemical Name or Material ACPT-I
Molecular Formula C8H11NO6
Assay Percent Range >98%
Synonym (1S,3R,4S)-1-Aminocyclopentane-1,3,4-tricarboxylic acid
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material ACPT-I
Assay Percent Range >98%
Molecular Formula C8H11NO6
Formula Weight 217.18
Synonym (1S,3R,4S)-1-Aminocyclopentane-1,3,4-tricarboxylic acid
Accessories

Desiccate at -20°C