Please login to your online account to display your discounted pricing

ACPT-I, Tocris

Group III mGlu agonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material ACPT-I
Assay Percent Range >98%
Molecular Formula C8H11NO6
Formula Weight 217.18
Synonym (1S,3R,4S)-1-Aminocyclopentane-1,3,4-tricarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

111110

 
r & d systems
RDS111110
10mg Each for $252.39

111150

 
r & d systems
RDS111150
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 217.18
Quantity 10mg
Chemical Name or Material ACPT-I
Synonym (1S,3R,4S)-1-Aminocyclopentane-1,3,4-tricarboxylic acid
Molecular Formula C8H11NO6
CAS 194918-76-8
Assay Percent Range >98%
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C