missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aconitine, 98%

$1,040.38 - $1,040.38

Chemical Identifiers

CAS 302-27-2
Molecular Formula C34H47NO11
Molecular Weight (g/mol) 645.746
MDL Number MFCD00082375
InChI Key XFSBVAOIAHNAPC-OZUFZMTGSA-N
Synonym aconitine
PubChem CID 132470875
SMILES CCN1CC2(C(CC(C34C2C(C(C31)C5(C6C4CC(C6OC(=O)C7=CC=CC=C7)(C(C5O)OC)O)OC(=O)C)OC)OC)O)COC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62056MD View Documents Alfa Aesar™
J62056MD
250mg Each for $1,040.38
 
Description

Description

  • Opens tetrodotoxin-sensitive sodium channels
  • Useful for creating models of cardiac arrhythmia
Specifications

Chemical Identifiers

302-27-2
645.746
XFSBVAOIAHNAPC-OZUFZMTGSA-N
132470875
C34H47NO11
MFCD00082375
aconitine
CCN1CC2(C(CC(C34C2C(C(C31)C5(C6C4CC(C6OC(=O)C7=CC=CC=C7)(C(C5O)OC)O)OC(=O)C)OC)OC)O)COC

Specifications

White
250mg
C34H47NO11
UN1544
14,118
Soluble in ethanol; Sparingly soluble in water
CCN1CC2(C(CC(C34C2C(C(C31)C5(C6C4CC(C6OC(=O)C7=CC=CC=C7)(C(C5O)OC)O)OC(=O)C)OC)OC)O)COC
132470875
Crystalline Powder
Aconitine
194°C to 202°C
302-27-2
MFCD00082375
74608
aconitine
XFSBVAOIAHNAPC-OZUFZMTGSA-N
645.746
645.75
98%
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c

H300+H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.

EINECSNumber : 206-121-7

RTECSNumber : AR5960000

TSCA : No

Recommended Storage : Store at -20°C