Learn More
Synbiosis™ aCOLyte 3 Validation Plate
The aCOLyte 3 Validation Plate checks the software and hardware performance for both pour and spiral plates. Test certificate included.
Supplier: Synbiosis™ ACOLVAL
Description
- Validates software and hardware performance for pour and spiral plates
- For use with aCOLyte 3 instruments
- Includes test certificate
Specifications
| Validation | |
| Synbiosis | |
| aCOLyte Instruments |
| Plastic | |
| Hardware and software validation for pour and spiral plates |