Please login to your online account to display your discounted pricing

Alfa Aesar™ Aclarubicin hydrochloride, 99%

Chemical Identifiers

Chemical Name or Material Aclarubicin hydrochloride
CAS 75443-99-1
Formula Weight 848.33
MDL Number MFCD00133024
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66842LB0

 
alfa aesar™
J66842LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 848.33
Melting Point 151° to 153°C
Chemical Name or Material Aclarubicin hydrochloride
Assay 99%
MDL Number MFCD00133024
Health Hazard 1 Danger
Physical Form Powder
Color Orange
Health Hazard 2 GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 75443-99-1
UN Number UN2811
Solubility Information Soluble in DMSO or DMF at 25mg/mL
Boiling Point 898°C
Quantity 5mg
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number QI9283500
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

2° to 8°C