Please login to your online account to display your discounted pricing

Acifran, Tocris

Hypolipidemic agent; agonist for the GPR109A (HM74A) and GPR109B receptors

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material Acifran
Assay Percent Range >99%
Molecular Formula C12H10O4
Formula Weight 218.21
Synonym 4, 5-Dihydro-5-methyl-4-oxo-5-phenyl-2-furancarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

176250

 
r & d systems
RDS176250
50mg Each for $746.48

176210

 
r & d systems
RDS176210
10mg Each for $177.37
Description & Specifications

Specifications

Formula Weight 218.21
Quantity 50mg
Chemical Name or Material Acifran
Assay Percent Range >99%
Molecular Formula C12H10O4
CAS 72420-38-3
Synonym 4, 5-Dihydro-5-methyl-4-oxo-5-phenyl-2-furancarboxylic acid
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C