Please login to your online account to display your discounted pricing

Acids Mix (GAC) (For EPA 604, 625)


Quantity 1x1mL
Packaging Ampule

Manufacturer: ultra scientific  EPA2008N1

 View more versions of this product

Catalog No. US-EPA-2008N1

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1x1mL
Methods For EPA Methods 604, 625
Chemical Name or Material Acids Mix (GAC)
Concentration or Composition (by Analyte or Components) Each component at 100μg/mL in Methanol:
4-Chloro-3-methylphenol
2-Chlorophenol
2,4-Dichlorophenol
2,4-Dimethylphenol
2,4-Dinitrophenol
4,6-Dinitro-2-methylphenol
2-Nitrophenol
4-Nitrophenol
Pentachlorophenol
Phenol
2,4,6-Trichlorophenol
Packaging Ampule

Specifications

Chemical Name or Material Acids Mix (GAC)