Please login to your online account to display your discounted pricing

Acids Mix (GAC) (For EPA 604, 625)

$113.71 - $113.71

Chemical Identifiers

Chemical Name or Material Acids Mix (GAC)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-EPA-2008N1

 
ultra scientific
EPA2008N1
Ampule 1x1mL Each for $113.71
Description & Specifications

Specifications

Quantity 1x1mL
Methods For EPA Methods 604, 625
Chemical Name or Material Acids Mix (GAC)
Concentration or Composition (by Analyte or Components) Each component at 100μg/mL in Methanol:
4-Chloro-3-methylphenol
2-Chlorophenol
2,4-Dichlorophenol
2,4-Dimethylphenol
2,4-Dinitrophenol
4,6-Dinitro-2-methylphenol
2-Nitrophenol
4-Nitrophenol
Pentachlorophenol
Phenol
2,4,6-Trichlorophenol
Packaging Ampule

Page 304C, Designated A2LA-Certified product.