missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acid Yellow 23, TCI America™

Manufacturer:  TCI America F0088500G

 View more versions of this product

Catalog No. F0088500G



Specifications

Specifications

Acid Yellow 23
500g
C16H9N4Na3O9S2
tartrazine, yellow 5, aizen tartrazine, trisodium salt, food yellow 4, tartrazine fd&c yellow #5, c.i. acid yellow 23, atul tartrazine, erio tartrazine, kako tartrazine
C1=CC(=CC=C1N=NC2C(=NN(C2=O)C3=CC=C(C=C3)S(=O)(=O)[O-])C(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+]
534.356
534.36
≥98.0% (HPLC)
Undesignated
1934-21-0
MFCD00148908
UJMBCXLDXJUMFB-UHFFFAOYSA-K
trisodium;5-oxo-1-(4-sulfonatophenyl)-4-[(4-sulfonatophenyl)diazenyl]-4H-pyrazole-3-carboxylate
164825
Crystal-Powder at 20°C

Chemical Identifiers

1934-21-0
534.356
UJMBCXLDXJUMFB-UHFFFAOYSA-K
164825
C1=CC(=CC=C1N=NC2C(=NN(C2=O)C3=CC=C(C=C3)S(=O)(=O)[O-])C(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+]
C16H9N4Na3O9S2
MFCD00148908
tartrazine, yellow 5, aizen tartrazine, trisodium salt, food yellow 4, tartrazine fd&c yellow #5, c.i. acid yellow 23, atul tartrazine, erio tartrazine, kako tartrazine
trisodium;5-oxo-1-(4-sulfonatophenyl)-4-[(4-sulfonatophenyl)diazenyl]-4H-pyrazole-3-carboxylate
Safety and Handling

Safety and Handling

EINECSNumber : (5)-1402

RTECSNumber : UQ6400000

TSCA : Yes

Documents