Please login to your online account to display your discounted pricing

Acid Surrogate Standard, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 5mL
Packaging 5mL amber prescored glass ampule

Manufacturer: spex certiprep llc  CLP90-SA5

 View more versions of this product

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. SPEX-N-023

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 5mL
Concentration or Composition (by Analyte or Components) 2000μg/mL
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
CAS 118-79-6
Analyte or Component Names 2,4,6-Tribromophenol
CAS 367-12-4
Solvent or Matrix Methanol
Health Hazard 1 Harmful if swallowed. Suspected of causing cancer.
Packaging 5mL amber prescored glass ampule
Chemical Name or Material Acid Surrogate Standard
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS 93951-73-6
Recommended Storage Ambient
DOT Information Hazardous
CAS 13127-88-3

Specifications

Chemical Name or Material Acid Surrogate Standard
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
2,4,6-Tribromophenol118-79-62000μg/mL
2-Chlorophenol-3,4,5,6-d493951-73-62000μg/mL
2-Fluorophenol367-12-42000μg/mL
Phenol-d613127-88-32000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Harmful if swallowed. Suspected of causing cancer.

Ambient