Please login to your online account to display your discounted pricing

Acid Surrogate Standard, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 1mL
Packaging 2mL amber prescored glass ampule

Manufacturer: spex certiprep llc  CLP90SA

 View more versions of this product

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. CLP90-SA

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1mL
Health Hazard 1 Harmful if swallowed. Suspected of causing cancer.
Chemical Name or Material Acid Surrogate Standard
Recommended Storage Ambient
Packaging 2mL amber prescored glass ampule
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
DOT Information Hazardous
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Analyte or Component Names 2,4,6-Tribromophenol
Solvent or Matrix Methanol
CAS 118-79-6
CAS 93951-73-6
Concentration or Composition (by Analyte or Components) 2000μg/mL
CAS 367-12-4
CAS 13127-88-3

Specifications

Chemical Name or Material Acid Surrogate Standard
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
2,4,6-Tribromophenol118-79-62000μg/mL
2-Chlorophenol-3,4,5,6-d493951-73-62000μg/mL
2-Fluorophenol367-12-42000μg/mL
Phenol-d613127-88-32000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Harmful if swallowed. Suspected of causing cancer.

Ambient