missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acid Surrogate Standard, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$176.77 - $310.57

Chemical Identifiers

For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Chemical Name or Material Acid Surrogate Standard
View More Specs

Disclaimers:

Product intended for laboratory use only


Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CLP90-SA Spex Certiprep Llc
CLP90SA
2mL amber prescored glass ampule 1mL Each for $310.57
CLP90-SA5 Spex Certiprep Llc
CLP90SA5
5mL amber prescored glass ampule 5mL Each for $176.77
SPEX-A-023 Spex Certiprep Llc
CLP90-SA
2mL amber prescored glass ampule 1mL Each for $310.57
SPEX-N-023 Spex Certiprep Llc
CLP90-SA5
5mL amber prescored glass ampule 5mL Each for $176.77
Specifications

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
2,4,6-Tribromophenol118-79-62000μg/mL
2-Chlorophenol-3,4,5,6-d493951-73-62000μg/mL
2-Fluorophenol367-12-42000μg/mL
Phenol-d613127-88-32000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Acid Surrogate Standard

Specifications

1mL
GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
2,4,6-Tribromophenol
118-79-6,93951-73-6,367-12-4,13127-88-3
Acid Surrogate Standard
2mL amber prescored glass ampule
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Methanol
2000μg/mL

Safety and Handling

Harmful if swallowed. Suspected of causing cancer.

DOTInformation : Hazardous

Recommended Storage : Ambient