missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acid Surrogate Standard, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC CLP90SA5
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
2,4,6-Tribromophenol | 118-79-6 | 2000μg/mL |
2-Chlorophenol-3,4,5,6-d4 | 93951-73-6 | 2000μg/mL |
2-Fluorophenol | 367-12-4 | 2000μg/mL |
Phenol-d6 | 13127-88-3 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Acid Surrogate Standard | |
5 mL | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
2000μg/mL | |
Methanol |
5mL amber prescored glass ampule | |
118-79-6,93951-73-6,367-12-4,13127-88-3 | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
2,4,6-Tribromophenol |
Product intended for laboratory use only