missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Acid Fuchsin
$125.35 - $348.12
Chemical Identifiers
CAS | 3244-88-0 |
---|---|
Molecular Formula | C20H17N3Na2O9S3 |
Molecular Weight (g/mol) | 587.544 |
MDL Number | MFCD00013286 |
InChI Key | VCEFFPMSOGDMJX-UHFFFAOYSA-N |
Synonym | Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S |
PubChem CID | 131852436 |
IUPAC Name | 3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium |
SMILES | CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC400210250
|
Thermo Scientific Chemicals
400210250 |
25 g | Glass bottle |
Each for $125.35
|
|
||||
AC400211000
|
Thermo Scientific Chemicals
400211000 |
100 g | Glass bottle |
Each for $348.12
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3244-88-0 | |
587.544 | |
VCEFFPMSOGDMJX-UHFFFAOYSA-N | |
131852436 | |
CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na] |
C20H17N3Na2O9S3 | |
MFCD00013286 | |
Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S | |
3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium |
Specifications
3244-88-0 | |
C20H17N3Na2O9S3 | |
15, 99 | |
Solubility in water: 143g/L. Other solubilities: slightly soluble in alcohol | |
CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na] | |
587.544 | |
585.52 | |
Glass bottle | |
25 g | |
Acid Fuchsin, Pure |
55% min. | |
MFCD00013286 | |
Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S | |
VCEFFPMSOGDMJX-UHFFFAOYSA-N | |
3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium | |
131852436 | |
Pure | |
Green | |
Glittering Powder |
RUO – Research Use Only