Please login to your online account to display your discounted pricing

Acid Detergent Solution, Fisher Chemical


Quantity 20L
Packaging PolyPac™

Manufacturer: fisher scientific company  SF11720

 View more versions of this product

Catalog No. SF11720

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 20L
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Colorless
Melting Point 0°C
Vapor Density >1.00g/mol
Concentration 0.995 to 1.005N
CAS 7732-18-5
ChemAlert Storage Symbol White
MDL Number MFCD00282010
Identification Pass Test
Packaging PolyPac™
CAS 7664-93-9
Chemical Name or Material Acid Detergent Solution
CAS 57-09-0
Concentration or Composition (by Analyte or Components) Cetyl Trimethylammonium Bromide (<5.0%), Sulfuric Acid (<5.5%), and Water (to balance)

Specifications

Chemical Name or Material Acid Detergent Solution
CAS 7664-93-9
CAS 57-09-0
CAS 7732-18-5
MDL Number MFCD00282010
Accessories
 

SDS & Safety Documents