Please login to your online account to display your discounted pricing

Acid Detergent Solution, Fisher Chemical

$287.38 - $971.08

Chemical Identifiers

Chemical Name or Material Acid Detergent Solution
CAS 7664-93-9
CAS 57-09-0
CAS 7732-18-5
MDL Number MFCD00282010
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SF1174

  SDS
fisher scientific company
SF1174
Poly Bottle 4L Each for $287.38
Case of 4 Each for $1,149.52

SF11720

  SDS
fisher scientific company
SF11720
PolyPac™ 20L Each for $971.08
Description & Specifications

Specifications

Quantity 4L
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Concentration 0.995 to 1.005N
Viscosity [NULL]
Color Colorless
Melting Point 0°C
Chemical Name or Material Acid Detergent Solution
Identification Pass Test
MDL Number MFCD00282010
ChemAlert Storage Symbol White
Packaging Poly Bottle
CAS 57-09-0
CAS 7732-18-5
CAS 7664-93-9
Concentration or Composition (by Analyte or Components) Cetyl Trimethylammonium Bromide (<5.0%), Sulfuric Acid (<5.5%), and Water (to balance)
Safety and Handling