missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acid Blue 25, MP Biomedicals™

Manufacturer:  Mp Biomedicals 15259525

 View more versions of this product

Catalog No. ICN15259525


 

Specifications Description & Specifications

Specifications

Acid Blue 25
Powder
416.4
C.I. 62055
Blue
6408-78-2
C20H13N2O5SNa
MFCD00001214
Solid
25g

Chemical Identifiers

6408-78-2
C20H13N2O5SNa
MFCD00001214
Acid Blue 25
Powder
416.4
C.I. 62055

Safety and Handling

EINECSNumber : 229-068-1

Recommended Storage : Store at Room Temperature (15° to 30°C)