Please login to your online account to display your discounted pricing

Acid Blue 25, MP Biomedicals™


Quantity 10g

Manufacturer: mp biomedicals  0215259510

 View more versions of this product

Catalog No. ICN15259510

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical State Solid
Quantity 10g
Chemical Name or Material Acid Blue 25
CAS 6408-78-2
Physical Form Blue powder
Molecular Formula C20H13N2O5SNa
Formula Weight 416.4g/mol
MDL Number MFCD00001214
Synonym C.I. 62055
Recommended Storage Store at Room Temperature (15° to 30°C)
EINECS Number 229-068-1

Specifications

Chemical Name or Material Acid Blue 25
CAS 6408-78-2
Physical Form Blue powder
Molecular Formula C20H13N2O5SNa
Formula Weight 416.4g/mol
MDL Number MFCD00001214
Synonym C.I. 62055

Stains and dyes

Accessories

Store at Room Temperature (15° to 30°C)