Please login to your online account to display your discounted pricing

Acid Blue 25, MP Biomedicals™

$87.93 - $87.93

Chemical Identifiers

Chemical Name or Material Acid Blue 25
CAS 6408-78-2
Physical Form Powder
Molecular Formula C20H13N2O5SNa
Formula Weight 416.4
MDL Number MFCD00001214
Synonym C.I. 62055
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15259525

 
mp biomedicals
15259525
25g Each for $87.93
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 416.4
Color Blue
Quantity 25g
EINECS Number 229-068-1
Physical State Solid
Chemical Name or Material Acid Blue 25
MDL Number MFCD00001214
Recommended Storage Store at Room Temperature (15° to 30°C)
Molecular Formula C20H13N2O5SNa
CAS 6408-78-2
Synonym C.I. 62055

Stains and dyes