missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acid Blank, 5% (v/v) Nitric Acid, Ricca Chemical

$157.18 - $243.79

Chemical Identifiers

CAS 7732-18-5
Physical Form Liquid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
PNACID5500 Ricca Chemical Company
RIPNACID5500
Poly Bottle 500mL Each for $157.18
PNACID51L Ricca Chemical Company
RIPNACID51L
Poly Bottle 1L Each for $243.79
Specifications Description & Specifications

Chemical Identifiers

7732-18-5
Liquid

Specifications

7732-18-5
Liquid
Poly Bottle
500mL
Clear colorless liquid
Odorless
Solubility in Water: Infinite
5%

Safety and Handling

Recommended Storage : Corrosive