missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acid Alcohol, Orth's Decolorizing Fluid, 3% HCl in 70% Alcohol, Ricca Chemical

$62.27 - $62.27

Chemical Identifiers

CAS 7732-18-5,64-17-5,67-56-1,7647-01-0
CAS Min % 26.0,66.0,3.0,2.9
CAS Max % 28.0,67.0,4.0,3.1
Physical Form Liquid
Chemical Name or Material Acid Alcohol, Orth's Decolorizing Flui
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
250-16 Ricca Chemical Company
RI25016
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
250-32 Ricca Chemical Company
RI25032
Poly natural 1L Each for $62.27
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,64-17-5,67-56-1,7647-01-0
28.0,67.0,4.0,3.1
Acid Alcohol, Orth's Decolorizing Flui
26.0,66.0,3.0,2.9
Liquid

Specifications

500mL
26.0,66.0,3.0,2.9
Liquid
3% HCl
Clear colorless
7732-18-5,64-17-5,67-56-1,7647-01-0
28.0,67.0,4.0,3.1
Poly natural
70% Alcohol
Acid Alcohol, Orth's Decolorizing Flui