Please login to your online account to display your discounted pricing

Acid-Alcohol, 3% (v/v) in 70% Alcohol, Certified, 3.0% (v/v) ±0.2%, LabChem™

$40.00 - $151.06

Chemical Identifiers

Chemical Name or Material Acid-Alcohol
CAS 64-17-5
CAS Max % 63
CAS 7732-18-5
CAS Max % 27
CAS 67-63-0
CAS Max % 3.5
CAS 67-56-1
CAS Max % 3.5
CAS 7647-01-0
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC105001

 
lab chem
LC105001
Each for $40.00

LC105004

 
lab chem
LC105004
Each for $151.06
Description & Specifications

Specifications

Assay 3.0% ± 0.2% (v/v)
Formula Weight 36.46
Purity Grade Certified Grade
CAS Max % 3.5
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide; May release flammable gases; Thermal decomposition generates: Corrosive vapors
Linear Formula HCl
Health Hazard 1 Danger
CAS Max % 3
CAS 64-17-5
CAS 7732-18-5
Identification Passes Test
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Causes serious skin burns and eye damage.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).
Physical Form Liquid
Packaging Poly Bottle
UN Number UN2924
CAS 67-63-0
Solubility Soluble in water
Color Clear, colorless
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 63
CAS 7647-01-0
Name Note 3% (v/v) in 70% Alcohol
Chemical Name or Material Acid-Alcohol
Recommended Storage Room Temperature
CAS 67-56-1
CAS Max % 27
CAS Max % 3.5
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Causes serious skin burns and eye damage.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).

Room Temperature