EVOS™ Achromat Oil Objectives
For general applications and image with different magnification
$917.71 - $1,101.35
Specifications
Product Type Achromat Oil Objective
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Focus Distance Magnification Power Numerical Aperture For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Focus Distance Magnification Power Numerical Aperture For Use With (Equipment) Price Quantity  
12-563-585 EVOS™
AMPFOP100
0.15mm 100X 1.25 EVOS™ XL Imaging System, EVOS FL Color Imaging System, EVOS FL Imaging System, EVOS FL Auto Imaging System, EVOS XL Core Imaging System Each for $917.71
12563584 EVOS™
AMPFOP050
0.19mm 50X 0.95 EVOS™ FL Color Imaging System, EVOS FL Auto Imaging System, EVOS FL Imaging System Each for $1,101.35
Specifications
Description
  • Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
  • Offers optical performance from visible light to near infrared light
  • Meets needs across spectrum of magnifications and optical specifications
Description & Specifications
Specifications
Achromat Oil Objective