missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Achatin-1

$90.07 - $129.54

Chemical Identifiers

CAS 121912-19-4
Molecular Formula C18H24N4O7
Molecular Weight (g/mol) 408.411
MDL Number MFCD00143539
InChI Key ZCPBEAHAVUJKAE-UHTWSYAYSA-N
Synonym achatin i, glycyl-phenylalanyl-alanyl-aspartic acid, gfad peptide, achatin-i, achatin-ii, gly-phe-ala-asp, gly-delta z-phe 2-ala-asp, l-aspartic acid, n-n-n-glycyl-d-phenylalanyl-l-alanyl, 2s-2-2s-2-2r-2-2-aminoacetyl amino-3-phenylpropanoyl amino propanoyl amino butanedioic acid
PubChem CID 5486682
IUPAC Name (2S)-2-[[(2S)-2-[[(2R)-2-[(2-aminoacetyl)amino]-3-phenylpropanoyl]amino]propanoyl]amino]butanedioic acid
SMILES CC(C(=O)NC(CC(=O)O)C(=O)O)NC(=O)C(CC1=CC=CC=C1)NC(=O)CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66486EXD View Documents Alfa Aesar™
J66486EXD
0.5mg Each for $90.07
 
AAJ66486MCR View Documents Alfa Aesar™
J66486MCR
1mg Each for $129.54
 
Specifications

Chemical Identifiers

121912-19-4
408.411
ZCPBEAHAVUJKAE-UHTWSYAYSA-N
5486682
CC(C(=O)NC(CC(=O)O)C(=O)O)NC(=O)C(CC1=CC=CC=C1)NC(=O)CN
C18H24N4O7
MFCD00143539
achatin i, glycyl-phenylalanyl-alanyl-aspartic acid, gfad peptide, achatin-i, achatin-ii, gly-phe-ala-asp, gly-delta z-phe 2-ala-asp, l-aspartic acid, n-n-n-glycyl-d-phenylalanyl-l-alanyl, 2s-2-2s-2-2r-2-2-aminoacetyl amino-3-phenylpropanoyl amino propanoyl amino butanedioic acid
(2S)-2-[[(2S)-2-[[(2R)-2-[(2-aminoacetyl)amino]-3-phenylpropanoyl]amino]propanoyl]amino]butanedioic acid

Specifications

White
121912-19-4
MFCD00143539
ZCPBEAHAVUJKAE-UHTWSYAYSA-N
(2S)-2-[[(2S)-2-[[(2R)-2-[(2-aminoacetyl)amino]-3-phenylpropanoyl]amino]propanoyl]amino]butanedioic acid
5486682
Solid
0.5mg
C18H24N4O7
achatin i, glycyl-phenylalanyl-alanyl-aspartic acid, gfad peptide, achatin-i, achatin-ii, gly-phe-ala-asp, gly-delta z-phe 2-ala-asp, l-aspartic acid, n-n-n-glycyl-d-phenylalanyl-l-alanyl, 2s-2-2s-2-2r-2-2-aminoacetyl amino-3-phenylpropanoyl amino propanoyl amino butanedioic acid
CC(C(=O)NC(CC(=O)O)C(=O)O)NC(=O)C(CC1=CC=CC=C1)NC(=O)CN
408.411
408.41
Achatin-1
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

Recommended Storage : −30° to −10°C