missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetylurea 99.0+%, TCI America™

$137.50 - $942.66

Chemical Identifiers

CAS 591-07-1
Molecular Formula C3H6N2O2
Molecular Weight (g/mol) 102.093
MDL Number MFCD00038140
InChI Key GKRZNOGGALENQJ-UHFFFAOYSA-N
Synonym acetylurea, acetamide, n-aminocarbonyl, n-acetylurea, acetylcarbamide, monoacetylurea, 1-acetylurea, acetic acid, ureide, n-aminocarbonyl acetamide, urea, acetyl, unii-8kru31i919
PubChem CID 68956
IUPAC Name N-carbamoylacetamide
SMILES CC(=O)NC(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A012425G View Documents TCI America
A012425G
25g Each for $137.50
 
A0124500G View Documents TCI America
A0124500G
500g Each for $942.66
 
Specifications

Chemical Identifiers

591-07-1
102.093
GKRZNOGGALENQJ-UHFFFAOYSA-N
68956
CC(=O)NC(=O)N
C3H6N2O2
MFCD00038140
acetylurea, acetamide, n-aminocarbonyl, n-acetylurea, acetylcarbamide, monoacetylurea, 1-acetylurea, acetic acid, ureide, n-aminocarbonyl acetamide, urea, acetyl, unii-8kru31i919
N-carbamoylacetamide

Specifications

White
25g
C3H6N2O2
acetylurea, acetamide, n-aminocarbonyl, n-acetylurea, acetylcarbamide, monoacetylurea, 1-acetylurea, acetic acid, ureide, n-aminocarbonyl acetamide, urea, acetyl, unii-8kru31i919
CC(=O)NC(=O)N
102.093
102.09
≥99.0% (N)
215°C
591-07-1
MFCD00038140
GKRZNOGGALENQJ-UHFFFAOYSA-N
N-carbamoylacetamide
68956
Crystal-Powder at 20°C
Acetylurea
Safety and Handling

Safety and Handling

RTECSNumber : AB4331500

TSCA : Yes