Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetylurea, 98%

$39.03 - $39.03

Chemical Identifiers

Chemical Name or Material Acetylurea
CAS 591-07-1
Molecular Formula C3H6N2O2
Formula Weight 102.09
MDL Number MFCD00038140
Beilstein 1751301
Synonym N-(Aminocarbonyl)acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0834306

  SDS
alfa aesar™
L0834306
5g Each for $39.03
Description & Specifications

Specifications

Formula Weight 102.09
Chemical Name or Material Acetylurea
Assay 98%
CAS 591-07-1
Synonym N-(Aminocarbonyl)acetamide
TSCA TSCA
Quantity 5g
RTECS Number AB4331500
Melting Point 214° to 218°C
Beilstein 1751301
Molecular Formula C3H6N2O2
MDL Number MFCD00038140