missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetylsalicylsalicylic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 269352500
Specifications
Acetylsalicylsalicylic acid | |
145°C to 160°C | |
97% | |
96.0 | |
96% min. (As is) Calculated (HPLC) | |
MFCD00143537 | |
14,1 | |
Solubility in water: .. Other solubilities: soluble in alkaline solutions,soluble in alcohol,practically insoluble in benzene | |
CC(=O)OC1=CC=CC=C1C(=O)OC2=CC=CC=C2C(=O)O | |
300.26 | |
300.26 |
Glass bottle | |
250mg | |
530-75-6 | |
100.0 | |
C16H12O6 | |
10,IV,165 | |
2-2-acetoxybenzoyl oxy benzoic acid, 2-carboxyphenyl 2-acetyloxy benzoate, acesalum, acetylsalicylsalicyic acid, acetylsalicylsalicylic acid, benzoic acid, 2-acetyloxy-, 2-carboxyphenyl ester, ccris 1740, diplosal acetate, diplosalsalate, unii-vbe72mcp5l | |
DDSFKIFGAPZBSR-UHFFFAOYSA-N | |
2-(2-acetyloxybenzoyl)oxybenzoic acid | |
10745 | |
97% |
Chemical Identifiers
530-75-6 | |
300.26 | |
DDSFKIFGAPZBSR-UHFFFAOYSA-N | |
10745 | |
CC(=O)OC1=CC=CC=C1C(=O)OC2=CC=CC=C2C(=O)O |
C16H12O6 | |
MFCD00143537 | |
2-2-acetoxybenzoyl oxy benzoic acid, 2-carboxyphenyl 2-acetyloxy benzoate, acesalum, acetylsalicylsalicyic acid, acetylsalicylsalicylic acid, benzoic acid, 2-acetyloxy-, 2-carboxyphenyl ester, ccris 1740, diplosal acetate, diplosalsalate, unii-vbe72mcp5l | |
2-(2-acetyloxybenzoyl)oxybenzoic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
If eye irritation persists: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
EINECSNumber : 208-493-6