missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetylpyrazine, 97%, Acros Organics

Manufacturer:  Fisher Scientific Acros 224660010

 View more versions of this product

Catalog No. AC224660010


 

Specifications Description & Specifications

Specifications

Acetylpyrazine
22047-25-2
100.0
Powder
97%
Glass bottle
MFCD00006134
Light yellow to yellow-brown
1g
97%
96.0
Authentic
≥96.0%
C6H6N2O
122.13
Methyl-2-pyrazinyl ketone
75° to 78°C

Chemical Identifiers

22047-25-2
100.0
C6H6N2O
MFCD00006134
Acetylpyrazine, 97%
96.0
97%
122.13
Methyl-2-pyrazinyl ketone
≥96.0%