Please login to your online account to display your discounted pricing

Acetylpyrazine, 97%, Acros Organics

$70.49 - $70.49

Chemical Identifiers

Chemical Name or Material Acetylpyrazine
Name Note 97%
CAS 22047-25-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H6N2O
Formula Weight 122.13
MDL Number MFCD00006134
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC224660010

  SDS
fisher scientific acros
224660010
Glass bottle 1g Each for $70.49
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 122.13
Physical Form Powder
Quantity 1g
Color Light yellow to yellow-brown
Melting Point 75° to 78°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material Acetylpyrazine
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
Molecular Formula C6H6N2O
Packaging Glass bottle
MDL Number MFCD00006134
CAS 22047-25-2
Synonym Methyl-2-pyrazinyl ketone