Please login to your online account to display your discounted pricing

Acetylenedicarboxylic Acid, Monopotassium salt 98+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC102530050

 View more versions of this product

Catalog No. AC102530050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.0%
Formula Weight 152.15
Quantity 5g
Color White
RTECS Number AP0700000
EINECS Number 213-169-2
TSCA TSCA
CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym 2-Butynedioic acid, monopotassium salt
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Acetylenedicarboxylic acid, monopotassium salt
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C4HKO4
Linear Formula HO2CCidentical toCCO2K
MDL Number MFCD00013143
CAS 928-04-1

Specifications

Chemical Name or Material Acetylenedicarboxylic acid, monopotassium salt
Name Note 98+%
CAS 928-04-1
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C4HKO4
Linear Formula HO2CCidentical toCCO2K
Formula Weight 152.15
MDL Number MFCD00013143
Synonym 2-Butynedioic acid, monopotassium salt
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.

SDS & Safety Documents