Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetylene-PEG4-sulforhodamine B conjugate

Chemical Identifiers

Chemical Name or Material Acetylene-PEG4-sulforhodamine B conjugate
Formula Weight 771.94
MDL Number MFCD22380745
Synonym Acetylene-Fluor 568
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64948LB0

 
alfa aesar™
J64948LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 771.94
Chemical Name or Material Acetylene-PEG4-sulforhodamine B conjugate
MDL Number MFCD22380745
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Solid
Synonym Acetylene-Fluor 568
Color Dark red
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5mg
Storage Note 1 -30° to -10°C

  • A sulforhodamine B based dye with acetylene functionality and a PEG spacer to improve solubility

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

-30° to -10°C