missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetylene-PEG4-biotin conjugate

An acetylene-biotin conjugate with PEG spacer

Chemical Identifiers

CAS 1262681-31-1
Molecular Formula C21H35N3O6S
Molecular Weight (g/mol) 457.59
MDL Number MFCD22380755
InChI Key SKMJWNZZFUDLKQ-UHFFFAOYNA-N
Synonym biotin-peg4-alkyne, acetylene-peg4-biotin conjugate
PubChem CID 75409985
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65577MB View Documents Alfa Aesar™
J65577MB
25mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ65577MC View Documents Alfa Aesar™
J65577MC
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1262681-31-1
457.59
SKMJWNZZFUDLKQ-UHFFFAOYNA-N
75409985
C21H35N3O6S
MFCD22380755
biotin-peg4-alkyne, acetylene-peg4-biotin conjugate

Specifications

White
1262681-31-1
MFCD22380755
SKMJWNZZFUDLKQ-UHFFFAOYNA-N
75409985
Solid
25mg
C21H35N3O6S
biotin-peg4-alkyne, acetylene-peg4-biotin conjugate
457.59
457.59
Acetylene-PEG4-biotin conjugate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C