Please login to your online account to display your discounted pricing

Acetylcholine iodide, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  0210002105

 View more versions of this product

Catalog No. ICN10002105

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 5g
Chemical Name or Material Acetylcholine iodide
CAS 2260-50-6
Physical Form White Powder
Assay Percent Range 0.991
Molecular Formula C7H16INO2
Formula Weight 273.11g/mol
Synonym Ach
Recommended Storage Store at 0°C.

Specifications

Chemical Name or Material Acetylcholine iodide
CAS 2260-50-6
Physical Form White Powder
Assay Percent Range 0.991
Molecular Formula C7H16INO2
Formula Weight 273.11g/mol
Synonym Ach
Accessories

Store at 0°C.