Please login to your online account to display your discounted pricing

Acetylcholine iodide, MP Biomedicals™

$55.53 - $55.53

Chemical Identifiers

Chemical Name or Material Acetylcholine iodide
CAS 2260-50-6
Physical Form White Powder
Assay Percent Range 0.991
Molecular Formula C7H16INO2
Formula Weight 273.11g/mol
Synonym Ach
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10002105

 
mp biomedicals
0210002105
5g Each for $55.53
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Acetylcholine iodide
CAS 2260-50-6
Physical Form White Powder
Assay Percent Range 0.991
Molecular Formula C7H16INO2
Formula Weight 273.11g/mol
Synonym Ach
Recommended Storage Store at 0°C.
Safety and Handling

Store at 0°C.