missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetylcholine iodide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1169414

 View more versions of this product

Catalog No. AAA1169414



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetylcholine iodide
161°C to 165°C
MFCD00011815
3571339
acetylcholine iodide, acetylcolina, 2-acetoxy-n,n,n-trimethylethanaminium iodide, acetylcolina italian, unii-7zcp12s7hq, choline, iodide, acetate ester, 2-acetoxyethyl trimethylammonium iodide, 7zcp12s7hq, ethanaminium, 2-acetyloxy-n,n,n-trimethyl-, iodide, 2-acetyloxy-n,n,n-trimethylethanamium iodide
[I-].CC(=O)OCC[N+](C)(C)C
273.11
273.11
2260-50-6
C7H16INO2
25 g
Hygroscopic; Light sensitive
SMBBQHHYSLHDHF-UHFFFAOYSA-M
2-acetyloxyethyl(trimethyl)azanium;iodide
75271
98%

Chemical Identifiers

2260-50-6
273.11
SMBBQHHYSLHDHF-UHFFFAOYSA-M
75271
[I-].CC(=O)OCC[N+](C)(C)C
C7H16INO2
MFCD00011815
acetylcholine iodide, acetylcolina, 2-acetoxy-n,n,n-trimethylethanaminium iodide, acetylcolina italian, unii-7zcp12s7hq, choline, iodide, acetate ester, 2-acetoxyethyl trimethylammonium iodide, 7zcp12s7hq, ethanaminium, 2-acetyloxy-n,n,n-trimethyl-, iodide, 2-acetyloxy-n,n,n-trimethylethanamium iodide
2-acetyloxyethyl(trimethyl)azanium;iodide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 218-862-3

RTECSNumber : KH3300000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only