Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetylated Peptide Certified Mass Spec Standard Mixture I


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67717LPL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67717LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Purification Method HPLC
Storage Requirements -30° to -10°C
Format Lyophilized powder
Regulatory Status RUO
Quantity 1 vial, 500 pmol
Sequence Angiotensin II, acetylated, Annexin I, acetylated, Calcitonin (15-29), acetylated
Product Type Acetylated Peptide Certified Mass Spec Standard Mixture I, 500pmol each peptide per vial 1 vial