Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetylated Calcitonin (15-29), Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67749LPL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67749LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Regulatory Status RUO
Sequence DFN-K(Ac)-FHTFPQTAIGV
Molecular Weight 1763.90
Format Lyophilized powder
Purification Method HPLC
Quantity 1 vial, 500 pmol
Product Type Acetylated Calcitonin (15-29), Certified Mass Spec Grade