missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetylacetone 99.0+%, TCI America™

Manufacturer:  TCI America P0052500ML

 View more versions of this product

Catalog No. P0052500ML



Specifications

Specifications

Acetylacetone
Yellow
500mL
C5H8O2
2310
YRKCREAYFQTBPV-UHFFFAOYSA-N
pentane-2,4-dione
31261
100.12
≥99.0% (GC)
138°C
-23°C
123-54-6
MFCD00008787
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
CC(=O)CC(=O)C
100.117
CHEBI:14750
Clear Liquid at 20°C

Chemical Identifiers

123-54-6
100.117
YRKCREAYFQTBPV-UHFFFAOYSA-N
31261
pentane-2,4-dione
C5H8O2
MFCD00008787
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
CHEBI:14750
CC(=O)CC(=O)C
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0562

RTECSNumber : SA1925000

TSCA : Yes

Documents