missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Riedel-de Haen™ Acetylacetone, puriss. p.a., ≥99.5% (GC), Honeywell™ Riedel-de-Haën™

Puriss. p.a.,≥99.5% (GC)

$121.74 - $121.74

Chemical Identifiers

CAS 123-54-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00008787
InChI Key YRKCREAYFQTBPV-UHFFFAOYSA-N
Synonym 2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
PubChem CID 31261
ChEBI CHEBI:14750
IUPAC Name pentane-2,4-dione
SMILES CC(=O)CC(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60-048-331 View Documents Honeywell Riedel-de Haen™
00900100ML
100mL Glass bottle Each for $121.74
 
Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications

Chemical Identifiers

123-54-6
100.117
YRKCREAYFQTBPV-UHFFFAOYSA-N
31261
pentane-2,4-dione
C5H8O2
MFCD00008787
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
CHEBI:14750
CC(=O)CC(=O)C

Specifications

Liquid
38°C (100.4°F)
1.452
139.5°C
100mL
C5H8O2
MFCD00008787
741937
Soluble in water
CC(=O)CC(=O)C
100.117
CHEBI:14750
≥99.5% (GC)
Acetylacetone
0.975g/mL at 25°C
Glass bottle
3.5 (vs air)
-23°C
123-54-6
CH3COCH2COCH3
UN2310
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
YRKCREAYFQTBPV-UHFFFAOYSA-N
pentane-2,4-dione
31261
100.12g/mol
puriss. p.a.
Safety and Handling

Safety and Handling

P261-P280-P311

H226-H302-H311-H331

DOTInformation : 6.1(3)/PGIII

EINECSNumber : 204-634-0

RTECSNumber : SA1925000