missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Acetylacetone, Reagent Grade, ≥99%, Honeywell™

Reagent Grade,≥99%

$68.85 - $387.40

Chemical Identifiers

CAS 123-54-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00008787
InChI Key YRKCREAYFQTBPV-UHFFFAOYSA-N
Synonym 2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
PubChem CID 31261
ChEBI CHEBI:14750
IUPAC Name pentane-2,4-dione
SMILES CC(=O)CC(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60048141 View Documents Honeywell™
P77541L
1L Glass bottle Each for $164.59
 
60048142 View Documents Honeywell™
P775425L
2.5L Glass bottle Each for $387.40
 
60048143 View Documents Honeywell™
P7754250ML
250mL Glass bottle Each for $68.85
 
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

123-54-6
100.117
YRKCREAYFQTBPV-UHFFFAOYSA-N
31261
pentane-2,4-dione
C5H8O2
MFCD00008787
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
CHEBI:14750
CC(=O)CC(=O)C

Specifications

Liquid
38°C (100.4°F)
Glass bottle
139.5°C (1013hPa)
-23°C
6 (at 200.00g/L)
C5H8O2
MFCD00008787
741937
153.8 - 155.2g/L at 20°C in water
CC(=O)CC(=O)C
100.117
CHEBI:14750
≥99%
0.975g/mL at 25°C
Characteristic
3.5 (vs air)
Colorless
1L
123-54-6
CH3COCH2COCH3
UN2310
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
YRKCREAYFQTBPV-UHFFFAOYSA-N
pentane-2,4-dione
31261
100.12g/mol
Acetylacetone
Safety and Handling

Safety and Handling

P210-P261-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378

DOTInformation : 6.1(3)/PGIII

RTECSNumber : 204-634-0