missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetylacetonato(1,5-cyclooctadiene)rhodium(I), 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC382545000

 View more versions of this product

Catalog No. AC382545000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Crystals
Yellow to orange
136° to 141°C
100.0
Acetylacetonato(1, 5-cyclooctadiene)rhodium(I)
Glass bottle
Authentic
12245-39-5
310.21
500mg
96.0
97%
C13 H19 O2 Rh
96.0% min. (ex Rh)
MFCD00075046

Chemical Identifiers

12245-39-5
100.0
C13 H19 O2 Rh
MFCD00075046
96.0
97%
310.21
Acetylacetonato(1, 5-cyclooctadiene)rhodium(I)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning