Please login to your online account to display your discounted pricing

Acetylacetonato(1,5-cyclooctadiene)rhodium(I), 97%, ACROS Organics™


Quantity 100mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC382541000

 View more versions of this product

Catalog No. AC382541000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 310.21
Physical Form Crystals
Color Yellow to orange
Quantity 100mg
Melting Point 136° to 141°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Acetylacetonato(1, 5-cyclooctadiene)rhodium(I)
Assay Percent Range 97%
CAS 12245-39-5
Molecular Formula C13 H19 O2 Rh
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable solid.
Packaging Glass bottle
Elemental Analysis 96.0% min. (ex Rh)
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00075046

Specifications

Chemical Name or Material Acetylacetonato(1, 5-cyclooctadiene)rhodium(I)
CAS 12245-39-5
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C13 H19 O2 Rh
Formula Weight 310.21
MDL Number MFCD00075046
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable solid.

SDS & Safety Documents