missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetylacetaldehyde dimethyl acetal, 92%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 292332500

 View more versions of this product

Catalog No. AC292332500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetylacetaldehyde dimethyl acetal
5436-21-5
100.0
Colorless to Yellow
178°C
Authentic
Glass bottle
1.4140 to 1.416
MFCD00008789
0.99
Solubility in water: decomposes. Other solubilities: miscible with most common organic solvents
COC(CC(C)=O)OC
132.16
1.503 mPa.s (20°C)
92%
95%
90.0
-82°C
0.9900g/mL
49°C
90% min. (GC)
C6H12O3
CH3COCH2CH(OCH3)2
250 mL
4,4-dimethoxy-2-butanone, acetylacetaldehyde dimethyl acetal, 2-butanone, 4,4-dimethoxy, 1,1-dimethoxy-3-butanone, acetoacetaldehyde dimethyl acetal, 3-oxobutyraldehyde dimethyl acetal, 4,4-dimethoxybutanone, acetylacetaldehyddimethylacetal, 3-ketobutyraldehyde dimethylacetal, formylacetone dimethyl acetal
PJCCSZUMZMCWSX-UHFFFAOYSA-N
4,4-dimethoxybutan-2-one
228548
132.16
Liquid

Chemical Identifiers

5436-21-5
132.16
PJCCSZUMZMCWSX-UHFFFAOYSA-N
228548
COC(CC(C)=O)OC
C6H12O3
MFCD00008789
4,4-dimethoxy-2-butanone, acetylacetaldehyde dimethyl acetal, 2-butanone, 4,4-dimethoxy, 1,1-dimethoxy-3-butanone, acetoacetaldehyde dimethyl acetal, 3-oxobutyraldehyde dimethyl acetal, 4,4-dimethoxybutanone, acetylacetaldehyddimethylacetal, 3-ketobutyraldehyde dimethylacetal, formylacetone dimethyl acetal
4,4-dimethoxybutan-2-one
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-605-1

RTECSNumber : EL7592500

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only