Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetyl-Neurotensin (8-13)


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66967MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66967MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Product Type Acetyl-Neurotensin (8-13)
Regulatory Status RUO
Purification Method HPLC
Sequence Ac-Arg-Arg-Pro-Tyr-Ile-Leu
Quantity 1mg
Molecular Weight 859.04
Format Solid