Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetyl-Myelin Basic Protein (4-14)Peptide

For cellular and molecular biology applications

Specifications

Product Type Acetyl-Myelin Basic Protein (4-14) Peptide
Molecular Weight 1432.70
Storage Requirements -30° to -10°C
Format Solid
Sequence N-Acetyl-Gln-Lys-Arg-Pro-Ser-Gln-Arg-Ser-Lys-Tyr-Leu
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66946EXD

 
alfa aesar™
J66946EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66946MCR

 
alfa aesar™
J66946MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Acetyl-Myelin Basic Protein (4-14) Peptide
Molecular Weight 1432.70
Storage Requirements -30° to -10°C
Format Solid
Sequence N-Acetyl-Gln-Lys-Arg-Pro-Ser-Gln-Arg-Ser-Lys-Tyr-Leu
Common Name Ac-MBP (4-14) Peptide
Regulatory Status RUO
Purification Method HPLC