Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetyl-Myelin Basic Protein (1-11), Human, Rat

For cellular and molecular biology applications

Specifications

Product Type Acetyl-Myelin Basic Protein (1-11), human, rat
Cross Reactivity Human, Rat
Molecular Weight 1293.42
Storage Requirements -30° to -10°C
Sequence N-Acetyl-Ala-Ser-Gln-Lys-Arg-Pro-Ser-Gln-Arg-His-Gly
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66603LB0

 
alfa aesar™
J66603LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66603MCR

 
alfa aesar™
J66603MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Acetyl-Myelin Basic Protein (1-11), human, rat
Cross Reactivity Human, Rat
Molecular Weight 1293.42
Storage Requirements -30° to -10°C
Sequence N-Acetyl-Ala-Ser-Gln-Lys-Arg-Pro-Ser-Gln-Arg-His-Gly
Format Solid
Regulatory Status RUO
Purification Method HPLC