Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetyl-Myelin Basic Protein (1-11), Human, Rat


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66603LB0

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66603LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 5mg
Format Solid
Sequence N-Acetyl-Ala-Ser-Gln-Lys-Arg-Pro-Ser-Gln-Arg-His-Gly
Product Type Acetyl-Myelin Basic Protein (1-11), human, rat
Molecular Weight 1293.42
Regulatory Status RUO
Storage Requirements -30° to -10°C
Cross Reactivity Human, Rat
Purification Method HPLC